Learn More
Bochem™ 18/10 Stainless Steel Double Spatula
Product Code. 10116460
Change view
Click to view available options
Length (Metric):
350 mm
400 mm
500 mm
Length (Metric) Blade:
85 mm
100 mm
Unit Size:
Each
Product Code. Length (Metric) Length (Metric) Blade unitSize
10116460 500 mm 100 mm Each
10720242 400 mm 100 mm Each
10094970 350 mm 85 mm Each
3 options
Product Code. 10116460
Brand: Bochem™ 3108
Only null left
Add to basket
Add to basket
Use the Bochem™ 8/10 Stainless Steel Double Spatula for many purposes in your laboratory..
  • Stainless Steel
  • Optional coatings available
  • Double Spatula
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Shape Straight
Product Type Double Ended Spatula
For Use With (Application) Multi-purpose spatula for a wide range of uses
Length (Metric) Blade 100 mm
Width (Metric) 20 mm
Width (Metric) Blade 20 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 6mm
Dimensions (L x W) 500 x 20 mm
Material Stainless Steel (18/10)
Length (Metric) 500 mm