Learn More
Bochem™ 18/10 Stainless Steel Double Spatula
Click to view available options
Length (Metric):
350 mm
400 mm
500 mm
Length (Metric) Blade:
85 mm
100 mm
Unit Size:
Each
Description
- Stainless Steel
- Optional coatings available
- Double Spatula
Specifications
Specifications
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |
| Shape | Straight |
| Product Type | Double Ended Spatula |
| For Use With (Application) | Multi-purpose spatula for a wide range of uses |
| Length (Metric) Blade | 85 mm |
| Width (Metric) | 16 mm |
| Width (Metric) Blade | 16 mm |
| Blade Material | Stainless Steel (18/10) |
| Diameter (Metric) Outer | 6mm |
| Show More |