Learn More
Bochem™ 18/10 Stainless Steel Double-Sided Chemical Spoons
Description
- Chemical spoon
- Double sided
- 18/10 stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Chemical Scoop |
| Handle Material | Stainless Steel (18/10) |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 250 mm |