Learn More
Bochem™ 18/10 Stainless Steel Coverglass Forceps
Product Code. 10323411
Change view
Click to view available options
Tip Style:
Bent
Straight
Unit Size:
Each
Pack of 10
Product Code. Tip Style unitSize
10323411 Bent Each
11825161 Straight Pack of 10
2 options
Product Code. 10323411
Brand: Bochem™ 1722
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Coverglass Forceps has 6mm wide points.
  • Coverglas forceps
  • Stainless steel
  • Points 6mm wide
Specifications
Tip Style Bent
Material Stainless Steel (18/10)
Length (Metric) 105 mm
Product Type Forceps
Quantity (No. per Case) 1 Pack
For Use With (Application) Good grip risk-free membranes, autoclaving or flame, pliers
Type Coverglass Forceps