Learn More
Bochem™ 18/10 Stainless Steel Chemical Spoon
Product Code. p-8000541
Change view
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
32 mm
40 mm
45 mm
48 mm
Diameter (Metric) Outer:
1 mm
2 mm
5 mm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) Length (Metric) Blade Diameter (Metric) Outer unitSize
10538881 150 mm 32 mm 5 mm Each
11865071 150 mm 32 mm 5 mm Pack of 10
10262091 150 mm 32 mm 1 mm Each
10485031 180 mm 32 mm 1 mm Each
10343981 210 mm 32 mm 1 mm Each
10373741 180 mm 40 mm 5 mm Each
11855071 210 mm 40 mm 5 mm Pack of 10
10169960 250 mm 45 mm 2 mm Each
10300681 250 mm 48 mm 5 mm Each
10613341 300 mm 48 mm 5 mm Each
10 options
Product Code. 10538881
Brand: Bochem™ 3461
Only null left
Add to basket
Add to basket
Stainless steel spoon for scooping chemicals.
  • Made of stainless steel
  • Double spoon
Specifications
Quantity 1 Ea.
Product Type Chemical Scoop
Length (Metric) Blade 32 mm
Width (Metric) 22 mm
Diameter (Metric) Outer 5 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm