Learn More
Bochem™ 18/10 E-Poli Silicone Beaker Tongs
Specifications
Specifications
| Length (Metric) Opening | 150 mm |
| Length (Metric) | 250 mm |
| Material | Stainless Steel (18/10) |
| Type | Beaker Tongs |