Learn More
Fisherbrand™ 150 Homogenizer Package
Product Code. 15585809 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
15585809 EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15585809
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 20-01-2027
Includes everything necessary to process samples as small as 200μL to 100mL. Fisherbrand™ 150 Homogenizer Package includes the 150w motor as well as accessories such as 2 saw tooth stainless steel generator probes, disposable generator probes and more.
  • Processes laboratory samples, from 200μL to 100mL
  • Powerful 150w motor
  • Lightweight ergonomic design
  • Quiet operation, under 72db
  • Available with disposable generator probes (adapter for use is included)
  • Cross-contamination can be completely eliminated with the use of plastic generator probes
  • Stainless steel generator probes are recommended for larger and tougher samples
Certifications
CE
Warranty
One year
TRUSTED_SUSTAINABILITY
Specifications
Speed 5000 to 35,000 rpm
Voltage 220 V
Electrical Requirements 220V EU Cord, 150W
Wattage 125 W
Volume Range (Metric) 200 μL to 100 mL
Diameter (Metric) 5.1 cm
Height (Metric) 16.5 cm
Certifications/Compliance CE, UKCA
For Use With (Application) Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Includes 150 W motor, Flat bottom Stainless Steel generator probe (5 x 75 mm), Saw tooth Stainless Steel generator probe (7 x 115 mm), Saw tooth Stainless Steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe adapter, Disposable generator probes, Instruction manual and Tool kit
Packaging Quantity 1 Ea.
Packaging Type Box
Plug Type EU Plug
Weight (Metric) 466 g
Product Type Homogenizer