Learn More
15-PGDH/HPGD Antibody (101), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90625PCP
4675.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18687820
  • 5755.00 NOK / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Mouse samples. It is validated for Western Blot, Immunoprecipitation