Learn More
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments
Product Code. p-4522214 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Ferrule
Packed Column
For Use With (Equipment):
Agilent Injectors and Non-MS Detectors
Agilent MS Detectors
Thermo Scientific MS Detectors
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Diameter (Metric):
0.1 to 0.25 mm
0.32 mm
0.53 mm
3.175 mm
6.35 mm
Unit Size:
Pack of 10
Product Code. Type For Use With (Equipment) Diameter (Metric) unitSize
10428874 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.32 mm Pack of 10
10478874 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.1 to 0.25 mm Pack of 10
10660046 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.53 mm Pack of 10
10466744 Ferrule Thermo Scientific MS Detectors 0.1 to 0.25 mm Pack of 10
10571115 Ferrule Thermo Scientific MS Detectors 0.32 mm Pack of 10
10117644 Ferrule Agilent MS Detectors 0.53 mm Pack of 10
10595615 Packed Column Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 6.35 mm Pack of 10
10554125 Packed Column Agilent Injectors and Non-MS Detectors 3.175 mm Pack of 10
10649485 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.1 to 0.25 mm Pack of 10
10630826 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.32 mm Pack of 10
10640826 Ferrule Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors 0.53 mm Pack of 10
11 options
Product Code. 10428874
Brand: Thermo Scientific™ 29033460
Only null left
Add to basket
Add to basket
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ Ferrules.
Use Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments to ensure optimal performance of your Thermo Scientific GC instrument. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Features
Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
Ferrules for different types of injectors and detectors
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description 29033460
Diameter (Metric) 0.32 mm
For Use With (Equipment) Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Material 15% Graphite, 85% Vespel