Learn More
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments to ensure optimal performance of your Thermo Scientific GC instrument. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
Ferrules for different types of injectors and detectors
Specifications
Specifications
| Type | Packed Column |
| Description | 290 VT165 |
| Diameter (English) | 1/4 in. |
| Diameter (Metric) | 6.35 mm |
| For Use With (Equipment) | Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors |
| Material | 15% Graphite, 85% Vespel |