Learn More
Keystone Cleanroom Products 1422A Tyvek™ Equipment Cover
GREENER_CHOICE
Product Code. 17745279 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W x H):
50 x 28 x 52 in.
52 x 50 x 52 in.
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
17745279 50 x 28 x 52 in. Pack of 50
17755279 52 x 50 x 52 in. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17745279 Supplier Keystone Cleanroom Products Supplier No. COVTVK502852SLT4
Only null left
Add to basket
Add to basket
Keystone Cleanroom Products Tyvek Autoclavable Covers are the ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
  • Keeps Cleaned Equipment Clean and Protected from Contamination
  • Breathable
  • Form Fitting, Fast and Consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type String Closure
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Thickness (English) 5.30 mil
Dimensions (L x W x H) 50 x 28 x 52 in.
Length (English) 50 in.
Width (English) 28 in.
Height (English) 52 in.
Height (Metric) 132 cm
Product Type Cover
Length (Metric) 127 cm
Width (Metric) 71.1 cm