Learn More
Keystone Cleanroom Products 1422A Tyvek™ Cylindrical Equipment Cover
GREENER_CHOICE
Product Code. 17796889 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Diameter (English):
14 in.
20 in.
23 in.
26 in.
32 in.
44 in.
48 in.
50 in.
Height (English):
5 in.
10 in.
20 in.
30 in.
36 in.
48 in.
60 in.
84 in.
Closure Type:
Elastic
Heat Seal
None
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. Diameter (English) Height (English) Closure Type unitSize
17796889 23 in. 5 in. Heat Seal Pack of 100
17758267 14 in. 60 in. None Pack of 50
17768267 14 in. 60 in. Elastic Pack of 50
17788267 20 in. 48 in. None Pack of 50
17798267 20 in. 48 in. Elastic Pack of 50
17871721 26 in. 20 in. Elastic Pack of 50
17778277 32 in. 30 in. None Pack of 50
17788277 32 in. 30 in. Elastic Pack of 50
17798277 32 in. 60 in. None Pack of 50
17778297 32 in. 60 in. Elastic Pack of 50
17788287 44 in. 10 in. Elastic Pack of 50
17748297 48 in. 36 in. None Pack of 50
17758297 48 in. 36 in. Elastic Pack of 50
17788297 48 in. 84 in. None Pack of 50
17798297 48 in. 84 in. Elastic Pack of 50
17738307 50 in. 60 in. None Pack of 25
17748307 50 in. 60 in. Elastic Pack of 25
17 missing translation for 'options'
Product Code. 17796889
missing translation for 'mfr': Keystone Cleanroom Products COVTVK0905E
Only null left
Add to basket
Add to basket
1422A Tyvek™ Cylindrical Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
  • Keeps cleaned equipment clean and protected from contamination
  • Breathable
  • Form fitting, fast and consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Heat Seal
Color White
Disposable Reusable
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 0.13 mm
Latex Free Yes
Protective Inner Packaging Poly Bag
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Thickness (English) 5.3 mil
Height (English) 5 in.
Height (Metric) 12.7 cm
Diameter (English) 23 in.
Diameter (Metric) 23 cm
Product Type Cover