Learn More
Keystone Cleanroom Products 1422A Tyvek™ Autoclavable Bowl Cover With Steam Indicator
GREENER_CHOICE
Product Code. 30238410 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Length (English):
32 in.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) unitSize
30238410 32 in. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30238410 Supplier Keystone Cleanroom Products Supplier No. BWLT2A32
Only null left
Add to basket
Add to basket
Autoclavable Bowl Cover prevents microbial and particulate contamination to critical parts.
  • Prevents microbial and particulate contamination to critical parts, while the fill line set-up activities are performed
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam Sterilization Indicator - Autoclavable at 121°C to 125°C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Specifications
Autoclavable Autoclavable
Certifications/Compliance CoC
Closure Type Elastic
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Temperature Range (Metric) 121°C to 125°C
Length (English) 32 in.
Width (English) 32 in.
Diameter (English) 21 to 27 in.
Diameter (Metric) 53 to 69 cm
Product Type Cover
Length (Metric) 81.28 cm
Width (Metric) 81.28 cm