Learn More
- Home
- Products
- Sterilizers and Autoclaves
- Autoclave Bags, Wraps, and Sleeves
- Keystone Cleanroom Products 1421B Tyvek™ Drawstring Closure Bag, 1 to 5 in. Wide
Keystone Cleanroom Products 1421B Tyvek™ Drawstring Closure Bag, 1 to 5 in. Wide
Product Code. 30020237
Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
2.5 x 3 in.
1 x 2 in.
3 x 2 in.
3 x 4 in.
4 x 2 in.
4 x 4 in.
4 x 9 in.
5 x 1 in.
5 x 2 in.
5 x 3 in.
5 x 5 in.
6 x 1 in.
6 x 2 in.
6 x 3 in.
6 x 5 in.
7 x 1 in.
7 x 2 in.
8 x 1 in.
8 x 2 in.
8 x 3 in.
8 x 5 in.
9 x 2 in.
9 x 3 in.
9 x 5 in.
10 x 2 in.
10 x 3 in.
10 x 4 in.
10 x 5 in.
11 x 1 in.
11 x 2 in.
11 x 5 in.
12 x 2 in.
12 x 3 in.
12 x 5 in.
13 x 2 in.
14 x 3 in.
15 x 5 in.
18 x 4 in.
25 x 4 in.
26 x 4 in.
28 x 4 in.
Unit Size:
Pack of 250
Pack of 500
41 product options available for selection
| Product Code. | Dimensions (L x W) | unitSize |
|---|---|---|
| 30020237 | 8 x 3 in. | Pack of 500 |
| 30020182 | 1 x 2 in. | Pack of 500 |
| 30020022 | 2.5 x 3 in. | Pack of 500 |
| 30020192 | 3 x 2 in. | Pack of 500 |
| 30020276 | 3 x 4 in. | Pack of 500 |
| 30020228 | 4 x 2 in. | Pack of 500 |
| 30020279 | 4 x 4 in. | Pack of 500 |
| 30020167 | 4 x 9 in. | Pack of 500 |
| 30020216 | 5 x 1 in. | Pack of 500 |
| 30020086 | 5 x 2 in. | Pack of 500 |
| 30020087 | 5 x 3 in. | Pack of 500 |
| 30020210 | 5 x 5 in. | Pack of 250 |
| 30020368 | 6 x 1 in. | Pack of 500 |
| 30020098 | 6 x 2 in. | Pack of 500 |
| 30020181 | 6 x 3 in. | Pack of 500 |
| 30020195 | 6 x 5 in. | Pack of 250 |
| 30020311 | 7 x 1 in. | Pack of 500 |
| 30020329 | 7 x 2 in. | Pack of 500 |
| 30020035 | 8 x 1 in. | Pack of 500 |
| 30020337 | 8 x 2 in. | Pack of 500 |
| 30020253 | 8 x 5 in. | Pack of 250 |
| 30019995 | 9 x 2 in. | Pack of 500 |
| 30020234 | 9 x 3 in. | Pack of 500 |
| 30020076 | 9 x 5 in. | Pack of 250 |
| 30020173 | 10 x 2 in. | Pack of 500 |
| 30020151 | 10 x 3 in. | Pack of 250 |
| 30020215 | 10 x 4 in. | Pack of 500 |
| 30020178 | 10 x 5 in. | Pack of 250 |
| 30020088 | 11 x 1 in. | Pack of 500 |
| 30020265 | 11 x 2 in. | Pack of 500 |
| 30020268 | 11 x 5 in. | Pack of 250 |
| 30020067 | 12 x 2 in. | Pack of 250 |
| 30020003 | 12 x 3 in. | Pack of 500 |
| 30020042 | 12 x 5 in. | Pack of 250 |
| 30020238 | 13 x 2 in. | Pack of 500 |
| 30020340 | 14 x 3 in. | Pack of 250 |
| 30020073 | 15 x 5 in. | Pack of 250 |
| 30020027 | 18 x 4 in. | Pack of 250 |
| 30020360 | 25 x 4 in. | Pack of 250 |
| 30020012 | 26 x 4 in. | Pack of 250 |
| 30020247 | 28 x 4 in. | Pack of 250 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 41 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams eliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Drawstring |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Steam Sterilization |
| For Use With (Equipment) | Pharmaceutical Equipment and Components |
| Material | Tyvek |
| Sterility | Non-sterile |
| Temperature Range (English) | Up to 250°F |
| Temperature Range (Metric) | Up to 125°C |
| Dimensions (L x W) | 8 x 3 in. |
| Length (English) | 8 in. |
| Width (English) | 3 in. |
| Product Type | Autoclave Bag |
| Capacity (English) | 24 oz. |
| Capacity (Metric) | 710 mL |
| Length (Metric) | 20 cm |
| Width (Metric) | 8 cm |
| Show More Show Less |