Learn More
Keystone Cleanroom Products 1421B Tyvek™ Drawstring Closure Bag, 1 to 5 in. Wide
GREENER_CHOICE
Product Code. 30020088 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
2.5 x 3 in.
1 x 2 in.
3 x 2 in.
3 x 4 in.
4 x 2 in.
4 x 4 in.
4 x 9 in.
5 x 1 in.
5 x 2 in.
5 x 3 in.
5 x 5 in.
6 x 1 in.
6 x 2 in.
6 x 3 in.
6 x 5 in.
7 x 1 in.
7 x 2 in.
8 x 1 in.
8 x 2 in.
8 x 3 in.
8 x 5 in.
9 x 2 in.
9 x 3 in.
9 x 5 in.
10 x 2 in.
10 x 3 in.
10 x 4 in.
10 x 5 in.
11 x 1 in.
11 x 2 in.
11 x 5 in.
12 x 2 in.
12 x 3 in.
12 x 5 in.
13 x 2 in.
14 x 3 in.
15 x 5 in.
18 x 4 in.
25 x 4 in.
26 x 4 in.
28 x 4 in.
Unit Size:
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) unitSize
30020088 11 x 1 in. Pack of 500
30020182 1 x 2 in. Pack of 500
30020022 2.5 x 3 in. Pack of 500
30020192 3 x 2 in. Pack of 500
30020276 3 x 4 in. Pack of 500
30020228 4 x 2 in. Pack of 500
30020279 4 x 4 in. Pack of 500
30020167 4 x 9 in. Pack of 500
30020216 5 x 1 in. Pack of 500
30020086 5 x 2 in. Pack of 500
30020087 5 x 3 in. Pack of 500
30020210 5 x 5 in. Pack of 250
30020368 6 x 1 in. Pack of 500
30020098 6 x 2 in. Pack of 500
30020181 6 x 3 in. Pack of 500
30020195 6 x 5 in. Pack of 250
30020311 7 x 1 in. Pack of 500
30020329 7 x 2 in. Pack of 500
30020035 8 x 1 in. Pack of 500
30020337 8 x 2 in. Pack of 500
30020237 8 x 3 in. Pack of 500
30020253 8 x 5 in. Pack of 250
30019995 9 x 2 in. Pack of 500
30020234 9 x 3 in. Pack of 500
30020076 9 x 5 in. Pack of 250
30020173 10 x 2 in. Pack of 500
30020151 10 x 3 in. Pack of 250
30020215 10 x 4 in. Pack of 500
30020178 10 x 5 in. Pack of 250
30020265 11 x 2 in. Pack of 500
30020268 11 x 5 in. Pack of 250
30020067 12 x 2 in. Pack of 250
30020003 12 x 3 in. Pack of 500
30020042 12 x 5 in. Pack of 250
30020238 13 x 2 in. Pack of 500
30020340 14 x 3 in. Pack of 250
30020073 15 x 5 in. Pack of 250
30020027 18 x 4 in. Pack of 250
30020360 25 x 4 in. Pack of 250
30020012 26 x 4 in. Pack of 250
30020247 28 x 4 in. Pack of 250
41 options
Product Code. 30020088
Brand: Keystone Cleanroom Products 6STK0111TD
Only null left
Add to basket
Add to basket
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Capacity (English) 11 oz.
Capacity (Metric) 325 mL
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Drawstring
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Material Tyvek
Sterility Non-sterile
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Dimensions (L x W) 11 x 1 in.
Length (English) 11 in.
Length (Metric) 28 cm
Width (English) 1 in.
Width (Metric) 3 cm
Product Type Autoclave Bag