Learn More
Keystone Cleanroom Products 1421B Tyvek™ Drawstring Closure Bag, 1 to 5 in. Wide
GREENER_CHOICE
Product Code. 30020022 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
2.5 x 3 in.
1 x 2 in.
3 x 2 in.
3 x 4 in.
4 x 2 in.
4 x 4 in.
4 x 9 in.
5 x 1 in.
5 x 2 in.
5 x 3 in.
5 x 5 in.
6 x 1 in.
6 x 2 in.
6 x 3 in.
6 x 5 in.
7 x 1 in.
7 x 2 in.
8 x 1 in.
8 x 2 in.
8 x 3 in.
8 x 5 in.
9 x 2 in.
9 x 3 in.
9 x 5 in.
10 x 2 in.
10 x 3 in.
10 x 4 in.
10 x 5 in.
11 x 1 in.
11 x 2 in.
11 x 5 in.
12 x 2 in.
12 x 3 in.
12 x 5 in.
13 x 2 in.
14 x 3 in.
15 x 5 in.
18 x 4 in.
25 x 4 in.
26 x 4 in.
28 x 4 in.
Unit Size:
Pack of 250
Pack of 500
Product Code. 30020022
Brand: Keystone Cleanroom Products 6STK02503TD
Only null left
Add to basket
Add to basket
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Drawstring
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Material Tyvek
Sterility Non-sterile
Quantity 500 Pk.
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Dimensions (L x W) 2.5 x 3 in.
Length (English) 2.5 in.
Length (Metric) 6.4 cm
Width (English) 3 in.
Width (Metric) 7.6 cm
Product Type Autoclave Bag