Learn More
Keystone Cleanroom Products 1421B Tyvek™ Autoclave Bag
GREENER_CHOICE
Product Code. 30020345 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
9.5 x 12 in.
12 x 2 in.
12 x 12 in.
15 x 10 in.
16 x 12 in.
18 x 12 in.
18 x 14 in.
20 x 20 in.
24 x 16 in.
24 x 20 in.
24 x 24 in.
26 x 24 in.
27 x 12 in.
28 x 44 in.
30 x 10 in.
30 x 18 in.
30 x 20 in.
32 x 12 in.
36 x 24 in.
36 x 30 in.
36 x 36 in.
40 x 12 in.
40 x 20 in.
42 x 24 in.
42 x 42 in.
46 x 12 in.
48 x 12 in.
48 x 24 in.
48 x 36 in.
48 x 48 in.
50 x 32 in.
54 x 50 in.
60 x 36 in.
62 x 10 in.
62 x 24 in.
Unit Size:
Pack of 100
Pack of 125
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) unitSize
30020345 28 x 44 in. Pack of 250
30020294 9.5 x 12 in. Pack of 250
30020126 12 x 2 in. Pack of 500
30020072 12 x 12 in. Pack of 250
30020064 15 x 10 in. Pack of 250
30020078 16 x 12 in. Pack of 250
30020106 18 x 12 in. Pack of 250
30020077 18 x 14 in. Pack of 250
30020090 20 x 20 in. Pack of 250
30020336 24 x 16 in. Pack of 250
30020250 24 x 20 in. Pack of 250
30020168 24 x 24 in. Pack of 250
30020130 26 x 24 in. Pack of 250
30020373 27 x 12 in. Pack of 250
30020346 30 x 10 in. Pack of 250
30020110 30 x 18 in. Pack of 250
30020351 30 x 20 in. Pack of 250
30020344 32 x 12 in. Pack of 250
30020053 36 x 24 in. Pack of 250
30019999 36 x 30 in. Pack of 250
30020260 36 x 36 in. Pack of 250
30020023 40 x 12 in. Pack of 250
30020049 40 x 20 in. Pack of 250
30020316 42 x 24 in. Pack of 250
30020267 42 x 42 in. Pack of 125
30020359 46 x 12 in. Pack of 250
30020032 48 x 12 in. Pack of 250
30020322 48 x 24 in. Pack of 250
30020282 48 x 36 in. Pack of 125
30020206 48 x 48 in. Pack of 125
30020246 50 x 32 in. Pack of 250
30020269 54 x 50 in. Pack of 100
30020157 60 x 36 in. Pack of 125
30020043 62 x 10 in. Pack of 250
30020079 62 x 24 in. Pack of 250
35 options
Product Code. 30020345
Brand: Keystone Cleanroom Products 6STK2844
Only null left
Add to basket
Add to basket
1421B Tyvek™ Autoclave Bag designed for the steam sterilization of large, or odd shaped pharmaceutical equipment change parts.
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment and Components
Material Tyvek
Sterility Non-sterile
Thickness (Metric) 0.13 mm
Quantity 250 Pk.
Temperature Range (English) Up to 250°F
Temperature Range (Metric) Up to 125°C
Thickness (English) 5.3 mil
Dimensions (L x W) 28 x 44 in.
Length (English) 28 in.
Length (Metric) 71.1 cm
Width (English) 44 in.
Width (Metric) 111.8 cm
Product Type Autoclave Bag