Learn More
Fisherbrand™ 14-Tooth Microplate Format Combs for Horizontal Midigel Systems
14-Tooth Microplate Format Combs for Horizontal Midigel Systems
Brand: Fisherbrand™
Additional Details : Weight : 0.90720kg
Description
- Accessory for use with Horizontal Midigel Systems
Specifications
Owl™ A1 Large Gel Electrophoresis System | |
Microplate | |
0.1 cm | |
0.28 in. |
Standard Comb | |
14 | |
35 μL | |
0.72 cm |