Description
- Tested by ISO 17025 accredited test method
- NIST traceable
- Low temperature coefficient of variation
- Guaranteed stability throughout entire shelf life even after opening bottle
- Accuracy: 1278 μS/cm ± 1%
Specifications
Specifications
| Packaging | HDPE bottle GL45 |
| Traceability to NIST | Yes |
| Physical Form | Liquid |
| For Use With (Equipment) | Conductivity Meter |
| Type | Conductivity Standard |
| Quantity | 500 mL |