Learn More
Watson-Marlow Pumps 120 Series Drive with Pumphead
Product Code. 17833084
Click to view available options
Motor Speed:
0.1 to 100 rpm
0.1 to 200 rpm
0.1 to 32 rpm
10 rpm
17 rpm
220 rpm
31 rpm
52 rpm
Pump Head:
102R
114DV
400D1
400DM2
400DM3
Model:
120F
120S
120U
Unit Size:
Each
Product Code. 17833084
Brand: Watson-Marlow Pumps 010.6112.R00
100 Series Pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.
101 series pumps are small and stackable lab pumps for handling valuable and delicate fluids without contamination. Save valuable space in cleanrooms, LAF and biosafety cabinets.
  • The 100 series is an ideal pump for maximizing yield in fermentation and cell culture applications.
  • Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimize your process.
  • The 100 series pump is suited to single-use systems.
  • Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate.
  • With a 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials.
  • A keypad lock secures your pumping parameters for consistent and repeatable performance.
Pumphead options available as standard (others available on request):
  • 120R single channel, two roller pumphead for continuous tubing
  • 114DV flip-top single-channel pumphead for continuous tubing
  • 400D1 single-channel pumphead for continuous tubing
  • 400DM2 twin-channel pumphead for manifold tubing
  • 400DM3 three-channel pumphead for manifold tubing
TRUSTED_SUSTAINABILITY
Specifications
Control Type Analog
Display Type LED
Enclosure Rating IP31
Flow Rate 0.003 to 54 mL/min.
Maximum Pressure 30 psi
Motor Speed 17 rpm
Model 120F
Pump Head 102R
Voltage 90/264 V