Learn More
BRAND™ 12-well Insert 2in1, PET-Membrane
Product Code. 16104967 Shop All Brand Products
Click to view available options
Pore Size:
0.4 μm
8 μm
No. per Pack:
4 x 9
48 x 1
Unit Size:
Pack of 36
Pack of 48
Product Code. 16104967
Brand: BRAND™ 782736
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
  • Use Inserts in a hanging or standing position
  • For 12-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Description 12-well Insert 2in1, PET-Membrane 8 μm
Material PET
Pore Size 8 μm
No. per Pack 48 x 1
Sterility Sterile
No. of Wells 12