Learn More
Megro™ 12-Ply Blue Bed Underlays, 40 x 60 cm
Product Code. 15839676
Change view
Click to view available options
Dimensions (L x W):
40 x 60 cm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15839676 40 x 60 cm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15839676 Supplier Megro™ Supplier No. MEG178604
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With a core made of recycling-cellulose with high suction power to contain liquids.
  • All around closed
  • Full extensive/skin-friendly/tear-proof fleece cover on the topside
  • Anti-skidding and waterproof foil underpart
  • Folded and ready to use
  • Provided 200 to a pack
Specifications
Product Type Underpad
Color Blue
Backsheet Material Foil
Topsheet Material Fleece
Dimensions (L x W) 40 x 60 cm
Format Folded
For Use With (Application) Keeping bedding dry
Ply 12-ply
Quantity 200/Pk.
Weight (Metric) 65 g