Learn More
Sartorius™ 11304 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters
11304 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters are hydrophilic, have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions (pH 4 – 8), hydrocarbons, and several other organic solvents.
Brand: Sartorius™ 11304--90------G
Additional Details : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- High, non-specific absorption
- Hydrophilic, with high flow rates
- Available in a range of pore sizes and diameters
- Ideal for several laboratory applications including particle retention, cell capture, cell retention, and cell filtration
Specifications
Autoclavable | |
White | |
Cellulose Nitrate | |
0.8 μm | |
200mL/min./cm2/bar | |
25/Pk | |
Membrane Filter | |
90 mm |
1.4bar | |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile | |
For many general laboratory applications where a membrane with a high non-specific adsorption is suitable | |
130 μm | |
90 mm |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.