Learn More
Hanna Instruments™ Chemical Test Kit Replacement Reagents
Click to view available options
For Use With (Application):
Acidity determination
Alkalinity determination
Carbon Dioxide determination
Chloride determination
Dissolved Oxygen determination
Free Chlorine determination
Iron determination
Phosphate determination
Salinity determination
Total Chlorine determination
Total hardness determination
No. of Tests:
50
100
110
Test Method:
Titration
Unit Size:
Each
Description
These high quality reagents are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability. Replacement titration reagent sets provide significant cost savings compared to purchasing an entirely new Test Kit.
Chemical Test Kit Replacement Reagents are available for the following Chemical Test Kits:
- Acidity Test Kit
- Alkalinity Test Kit
- Ascorbic Acid Test Kit
- Cardon Dioxide Test Kit
- Chloride Test Kit
- Dissolved Oxgen Test Kit
- Free and Total Chlorine (Low, Medium, High Range) Test Kit
- Free Chlorine Test Kit
- Total Chlorine Test Kit
- Iron (Medium Range) Test Kit
- Phosphate Test Kit
- Salinity Test Kit
- Sulfite Test Kit
- Total Hardness Test Kit
Specifications
Specifications
| Analytes | Mercuric nitrate |
| For Use With (Application) | Salinity determination |
| For Use With (Equipment) | HI 3835 Salinity Chemical Test Kit |
| Includes | Bottles with Protective Cap and Instructions |
| No. of Tests | 110 |
| Quantity | 1 Set |
| Range | 0.0 to 40.0g/kg (ppt) |
| Test Method | Titration |
| Type | Reagent Set |