missing translation for 'onlineSavingsMsg'
Learn More

11-Chloro-1-undecene, 97%

Product Code. 15400278
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 15400278

Brand: Thermo Scientific Alfa Aesar H53449.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 872-17-3 | C11H21Cl | 188.74 g/mol

11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-17-3
Molecular Formula C11H21Cl
Molecular Weight (g/mol) 188.74
MDL Number MFCD00671354
InChI Key PSEVKFKRYVAODC-UHFFFAOYSA-N
Synonym 11-chloro-1-undecene, 1-undecene, 11-chloro, 10-undecenyl chloride, undecylenyl chloride, acmc-1bjnc, undec-10-enyl chloride, 11-chloro-1-undecene #
PubChem CID 543805
IUPAC Name 11-chloroundec-1-ene
SMILES ClCCCCCCCCCC=C

Specifications

Density 0.995
Boiling Point 108°C to 109°C (6 mmHg)
Flash Point 104°C (219°F)
Linear Formula Cl(CH2)9CH=CH2
Refractive Index 1.451
Quantity 25 mL
Solubility Information Soluble in water.
Formula Weight 188.74
Percent Purity 97%
Chemical Name or Material 11-Chloro-1-undecene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.