Learn More
11β,17-Dihydroxypregn-4-ene-3,20-dione (Oxenol), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
11β,17-Dihydroxypregn-4-ene-3,20-dione (Oxenol)
Specifications
Specifications
| Chemical Name or Material | 21-Deoxycortisol |
| CAS | 641-77-0 |
| Analyte or Component Names | 21-Deoxycortisol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H30 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pregn-4-ene-3,20-dione, 11,17-dihydroxy-, (11β)-, Pregn-4-ene-3,20-dione, 11β,17-dihydroxy- (8CI), Progesterone, 11β,17-dihydroxy- (6CI), 11β,17α-Dihydroxypregn-4-ene-3,20-dione, 11β,17α-Dihydroxyprogesterone, 21-Dehydrohydrocortisone, 21-Deoxycortisol, 21-Deoxyhydrocortisone, 21-Desoxycortisol, Pregn-4-ene-11β,17α-diol-3,20-dione |
| InChI Formula | InChI=1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1 |
| SMILES | CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C |
| IUPAC Name | (8S,9S,10R,11S,13S,14S,17R)-17-acetyl-11,17-dihydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido