Learn More
Biochrom Quartz Flowcell with Screw and Connectors
Product Code. 11683024 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Flow Cell
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11683024 Flow Cell Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11683024 Supplier Biochrom Supplier No. 80710040
  • ONLINE EXCLUSIVE PRICE
    9730.00 NOK
    8937.07 NOK / Pack of 1
    Save 792.93 NOK
    8% Off
Estimated Shipment: 13-01-2027
Quartz Flowcell with Screw and Connectors for use with Sipper Accessory of Ultrospec 7000/8000/9000 UV-Vis Spectrophotometers.
  • Easy to assemble
Specifications
Type Flow Cell
Description 10 mm Quartz Flowcell with Screw and Connectors for Sipper Accessory
Quantity 1 Ea.
Pathlength 10 mm
Volume (Metric) 80 μL
For research use only.