Learn More
R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X)
Description
- Maintains healthy cardiomyocyte population for longer than 200 days
- High lot-to-lot consistency decreases experimental variation
Repeated mechanical stress can negatively affect cell attachment and survival. For extended culture, daily media exchanges should be handled as gently as possible.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Concentration | 50 X |
| Form | Liquid |
| Product Type | Maintenance Media Supplement |
| For Use With (Application) | Detection of Cardiac Troponin T in Human ES-derived Cardiomyocytes, Troponin I Expression in Human ES-derived Cardiomyocytes |
| Quantity | 10 mL |
| Regulatory Status | RUO |