missing translation for 'onlineSavingsMsg'
Learn More

Z-Ile-Leu-aldehyde, MedChemExpress

Product Code. 16415188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16415188 10 mg 10mg
16467907 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16415188

Brand: Medchem Express HY12465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Z-Ile-Leu-aldehyde (Z-IL-CHO) is a potent and competitive peptide aldehyde inhibitor of γ-secretase and notch.

Z-Ile-Leu-aldehyde (Z-IL-CHO) is a potent and competitive peptide aldehyde inhibitor of γ-secretase and notch.

Specifications

Chemical Name or Material Z-Ile-Leu-aldehyde
Physical Form Solid
CAS 161710-10-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H30N2O4
For Use With (Application) Cancer-programmed cell death
Synonym Z-IL-CHO GSI-XII γ-Secretase inhibitor XII
Solubility Information DMSO : ≥ 41 mg/mL (113.12 mM)
SMILES O=C(OCC1=CC=CC=C1)N[C@H](C(N[C@H](C=O)CC(C)C)=O)[C@@H](C)CC
Molecular Weight (g/mol) 362.46
Formula Weight 362.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.