missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Capsaicin, MedChemExpress

Product Code. 16498708
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498708

Brand: Medchem Express HYB1583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain.

(Z)-Capsaicin is the cis isomer of capsaicin, acts as an orally active TRPV1 agonist, and is used in the research of neuropathic pain.

Specifications

Chemical Name or Material (Z)-Capsaicin
CAS 25775-90-0
Color White
Purity Grade Notes Research
Molecular Formula C18H27NO3
Quantity 10 mg
Synonym Zucapsaicin Civamide cis-Capsaicin
Solubility Information DMSO : ≥ 125 mg/mL (409.29 mM)
SMILES CC(C)/C=C\CCCCC(NCC1=CC=C(O)C(OC)=C1)=O
Molecular Weight (g/mol) 305.41
Formula Weight 305.41
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.