missing translation for 'onlineSavingsMsg'
Learn More

Toltrazuril (sulfone), MedChemExpress

Product Code. 16407268
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16407268

Brand: Medchem Express HY1700810mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril (HY-B0175), with antiprotozoal activity. Toltrazuril sulfone is a triazine anticoccidial that is developed to prevent coccidiosis in poultry.

Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril (HY-B0175), with antiprotozoal activity. Toltrazuril sulfone is a triazine anticoccidial that is developed to prevent coccidiosis in poultry.

Specifications

Chemical Name or Material Toltrazuril (sulfone)
Physical Form Solid
CAS 69004-04-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H14F3N3O6S
For Use With (Application) COVID-19-immunoregulation
Synonym Ponazuril
Solubility Information DMSO : 50 mg/mL (109.32 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C=C1)OC2=C(C=C(C=C2)N3C(N(C(NC3=O)=O)C)=O)C)(C(F)(F)F)=O
Molecular Weight (g/mol) 457.38
Formula Weight 457.38
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.