missing translation for 'onlineSavingsMsg'
Learn More

Talipexole dihydrochloride, MedChemExpress

Product Code. 16468258
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468258 10 mg 10mg
16444147 50 mg 50mg
16407986 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468258 Supplier Medchem Express Supplier No. HYA0008
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Talipexole dihydrochloride (B-HT 920 dihydrochloride) is a dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist, which displays antiParkinsonian activity.

Talipexole dihydrochloride (B-HT 920 dihydrochloride) is a dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist, which displays antiParkinsonian activity.

Specifications

Chemical Name or Material Talipexole dihydrochloride
Physical Form Solid
CAS 36085-73-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H17Cl2N3S
For Use With (Application) Neuroscience-Neurodegeneration
Synonym B-HT 920 dihydrochloride
Solubility Information DMSO : 28 mg/mL (99.21 mM; Need ultrasonic and warming)
SMILES NC1=NC2=C(CCN(CC2)CC=C)S1.Cl.Cl
Molecular Weight (g/mol) 282.23
Formula Weight 282.23
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.