missing translation for 'onlineSavingsMsg'
Learn More

SU9516, MedChemExpress

Product Code. 16457548
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457548 10 mg 10mg
16473567 50 mg 50mg
16420028 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457548 Supplier Medchem Express Supplier No. HY18629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SU9516 is a potent CDK2 inhibitor, with an IC50 of 22 nM, and also shows inhibitory effects on CDK1 and CDK4, with IC50s of 40, 200 nM, respectively.

SU9516 is a potent CDK2 inhibitor, with an IC50 of 22 nM, and also shows inhibitory effects on CDK1 and CDK4, with IC50s of 40, 200 nM, respectively.

Specifications

Chemical Name or Material SU9516
CAS 377090-84-1
Color Brown
Purity Grade Notes Research
Molecular Formula C13H11N3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (414.51 mM)
SMILES O=C1NC2=C(C=C(OC)C=C2)/C1=C/C3=CN=CN3
Molecular Weight (g/mol) 241.25
Formula Weight 241.25
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.