missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR131675, MedChemExpress
Description
SAR131675 is a potent and selective VEGFR3 inhibitor with an IC50 of 23 nM.
Specifications
Specifications
| Chemical Name or Material | SAR131675 |
| CAS | 1433953-83-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H22N4O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 100 mg/mL (279.03 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C1C(C(NC)=O)=C(N)N(CC)C2=NC(C#C[C@@](COC)(C)O)=CC=C21 |
| Molecular Weight (g/mol) | 358.39 |
| Formula Weight | 358.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?