Learn More
MedChemExpress Porcine dynorphin A(1-13)
Product Code. 16439358
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 16439358
Brand: MedChemExpress HYP0088
MedChemExpress Porcine dynorphin A (1-13) is a potent, endogenous κ opioid receptor agonist and is antinociceptive at physiological concentrations.
Specifications
Purity 99.61%
CAS 72957-38-1
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1603.96 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 60 mg/mL (37.41 mM)
Synonym Dynorphin A Porcine Fragment 1-13
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.