missing translation for 'onlineSavingsMsg'
Learn More

Otilonium bromide, MedChemExpress

Product Code. 16448508
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16448508

Brand: Medchem Express HYB0499A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Octylonium bromide (SP63) is an antimuscarinic used as a spasmolytic agent.

Octylonium bromide (SP63) is an antimuscarinic used as a spasmolytic agent.

Specifications

Chemical Name or Material Otilonium bromide
CAS 26095-59-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H43BrN2O4
Quantity 10 mg
Synonym Octylonium bromide SP63
Solubility Information H2O : ≥ 100 mg/mL (177.44 mM) ∣DMSO : 50 mg/mL (88.72 mM; Need ultrasonic)
SMILES O=C(OCC[N+](CC)(CC)C)C1=CC=C(NC(C2=CC=CC=C2OCCCCCCCC)=O)C=C1.[Br-]
Molecular Weight (g/mol) 563.57
Formula Weight 563.57
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.