missing translation for 'onlineSavingsMsg'
Learn More

Nestoron, MedChemExpress

Product Code. 16435538
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16435538

Brand: Medchem Express HY13071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nestoron (ST-1435) is a 19-norprogesterone derivative with high affinity and selectivity for progesterone receptors. Nestoron is a highly selective and potent progestogen that can be used as a hormonal contraceptive.

Nestoron (ST-1435) is a 19-norprogesterone derivative with high affinity and selectivity for progesterone receptors. Nestoron is a highly selective and potent progestogen that can be used as a hormonal contraceptive.

Specifications

Chemical Name or Material Nestoron
Physical Form Solid
CAS 7759-35-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H30O4
Synonym ST-1435 Elcometrine
Solubility Information DMSO : 33.33 mg/mL (89.96 mM; Need ultrasonic)
SMILES CC([C@]1(OC(C)=O)[C@@]2([C@]([H])([C@@]3(CCC4=CC(CC[C@@]4([C@]3(CC2)[H])[H])=O)[H])CC1=C)C)=O
Molecular Weight (g/mol) 370.48
Formula Weight 370.48
Percent Purity 99.84%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.