missing translation for 'onlineSavingsMsg'
Learn More

Letrozole, TRC

Product Code. 30011722
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011722 10 mg 10mg
30134964 25 mg 25mg
30133252 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011722 Supplier Toronto Research Chemicals Supplier No. TRCL33010010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112809-51-5
Molecular Formula C17 H11 N5
Molecular Weight (g/mol) 285.3
Synonym Letrozole, Benzonitrile, 4,4'-(1H-1,2,4-triazol-1-ylmethylene)bis-, 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)bis[benzonitrile], 4,4'-(1H-1,2,4-Triazol-1-ylmethylene)dibenzonitrile, 4-[1-(4-Cyanophenyl)-1-(1,2,4-triazol-1-yl)methyl]benzonitrile, CGS 20267, Femara, Lerozole, Letocor, Letoripe, Letrazole, Letrof L Lil, Letroz
IUPAC Name 4-[(4-cyanophenyl)-(1,2,4-triazol-1-yl)methyl]benzonitrile
SMILES N#Cc1ccc(cc1)C(c2ccc(cc2)C#N)n3cncn3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H
Formula Weight 285.1014
Percent Purity >95%
Chemical Name or Material Letrozole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.