missing translation for 'onlineSavingsMsg'
Learn More

GW788388, MedChemExpress

Product Code. 16443818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16443818 10 mg 10mg
16444866 100 mg 100mg
16420197 50 mg 50mg
16406547 5 mg 5mg
4 options
This item is not returnable. View return policy

Product Code. 16443818

Brand: Medchem Express HY10326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GW788388 is a potent and selective inhibitor of ALK5 with IC50 of 18 nM, and also inhibits TGF-β type II receptor and activin type II receptor activities, without inhibiting BMP type II receptor.

GW788388 is a potent and selective inhibitor of ALK5 with IC50 of 18 nM, and also inhibits TGF-β type II receptor and activin type II receptor activities, without inhibiting BMP type II receptor.

Specifications

Chemical Name or Material GW788388
CAS 452342-67-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H23N5O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (58.76 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1)C2=NC=CC(C3=CNN=C3C4=NC=CC=C4)=C2)NC5CCOCC5
Molecular Weight (g/mol) 425.48
Formula Weight 425.48
Percent Purity 98.3%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.