missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNE-617 hydrochloride, MedChemExpress
Description
GNE-617 hydrochloride is a specific NAMPT inhibitor that inhibits the biochemical activity of NAMPT with an IC50 of 5 nM and exhibits efficacy in xenograft models of cancer.
Specifications
Specifications
| Chemical Name or Material | GNE-617 hydrochloride |
| CAS | 2070014-99-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H16ClF2N3O3S |
| Quantity | 10 mg |
| Solubility Information | DMSO : 5.56 mg/mL (11.99 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(NCC1=CC=C(S(C2=CC(F)=CC(F)=C2)(=O)=O)C=C1)C3=CN4C(C=C3)=NC=C4.Cl |
| Molecular Weight (g/mol) | 463.88 |
| Formula Weight | 463.88 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?