missing translation for 'onlineSavingsMsg'
Learn More

Edoxaban tosylate, MedChemExpress

Product Code. 16403758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16403758 10 mg 10mg
16466477 5 mg 5mg
16464816 100 mg 100mg
16440137 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16403758

Brand: Medchem Express HY10264A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Edoxaban (DU-176b) tosylate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban tosylate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban tosylate can be used for preventing thromboembolic disease research.

Edoxaban (DU-176b) tosylate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban tosylate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban tosylate can be used for preventing thromboembolic disease research.

Specifications

Chemical Name or Material Edoxaban tosylate
CAS 480449-71-6
Color White
Purity Grade Notes Research
Molecular Formula C31H38ClN7O7S2
Quantity 10 mg
Synonym DU-176b
Solubility Information DMSO : 100 mg/mL (138.84 mM; Need ultrasonic)
SMILES O=C(N(C)C)[C@@H](CC1)C[C@H]([C@H]1NC(C(NC(C=C2)=NC=C2Cl)=O)=O)NC(C3=NC(CC4)=C(CN4C)S3)=O.O=S(C5=CC=C(C)C=C5)(O)=O
Molecular Weight (g/mol) 720.26
Formula Weight 720.26
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.