missing translation for 'onlineSavingsMsg'
Learn More

CP21R7, MedChemExpress

Product Code. 16412848
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412848

Brand: Medchem Express HY100207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CP21R7 is potent GSK-3β inhibitor, with an IC50 of 1.8 nM; CP21R7 also shows inhibitory activitiy against PKCα, with an IC50 of 1900 nM.

CP21R7 is potent GSK-3β inhibitor, with an IC50 of 1.8 nM

Specifications

Chemical Name or Material CP21R7
CAS 125314-13-8
Color Orange
Purity Grade Notes Research
Molecular Formula C19H15N3O2
Quantity 10 mg
Synonym CP21
Solubility Information DMSO : ≥ 32 mg/mL (100.84 mM)
SMILES O=C1C(C2=CN(C)C3=CC=CC=C23)=C(C4=CC(N)=CC=C4)C(N1)=O
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.