missing translation for 'onlineSavingsMsg'
Learn More

CP-673451, MedChemExpress

Product Code. 16424958
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424958

Brand: Medchem Express HY12050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively.

CP-673451 is a potent and selective inhibitor of PDGFR with IC50s of 10 and 1 nM for PDGFRα and PDGFRβ, respectively.

Specifications

Chemical Name or Material CP-673451
CAS 343787-29-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H27N5O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (239.52 mM)
SMILES NC1CCN(C2=C3N=C(C=CC3=CC=C2)N4C=NC5=CC(OCCOC)=CC=C54)CC1
Molecular Weight (g/mol) 417.5
Formula Weight 417.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.