missing translation for 'onlineSavingsMsg'
Learn More

cFMS Receptor Inhibitor II, MedChemExpress

Product Code. 16434638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434638 10 mg 10mg
16497347 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16434638 Supplier Medchem Express Supplier No. HY112451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine.

cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine.

Specifications

Chemical Name or Material cFMS Receptor Inhibitor II
CAS 959860-85-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H20N4O
Quantity 10 mg
Solubility Information DMSO : 83.33 mg/mL (226.18 mM; Need ultrasonic)
SMILES O=C(C1=C(NC2=CC=C(C)C(C)=C2)C3=CC=C(C4=CC=NC=C4)C=C3N=C1)N
Molecular Weight (g/mol) 368.43
Formula Weight 368.43
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.