Learn More
Cefditoren (Pivoxil), MedChemExpress
Product Code. 16477348
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Product Code. Quantity unitSize
16477348 10 mg 10mg
16483367 50 mg 50mg
16447316 100 mg 100mg
16421266 200 mg 200mg
4 options
Product Code. 16477348
Brand: Medchem Express HY17452A
MedChemExpress Cefditoren Pivoxil (ME 1207) is a broad-spectrum, third-generation, oral cephalosporin antibacterial with enhanced stability against many common β lactamases.
Cefditoren Pivoxil has activity against Gram-negative organisms and Gram-positive organisms. Cefditoren Pivoxil can be used in the research of infection diseases such as acute exacerbations of chronic bronchitis, community-acquired pneumonia (CAP), streptococcal pharyngitis/tonsillitis, or uncomplicated skin and skin structure infections.
Specifications
Chemical Name or Material Cefditoren (Pivoxil)
Physical Form Solid
CAS 117467-28-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H28N6O7S3
For Use With (Application) COVID-19-immunoregulation
Synonym Cefditoren pivoxyl Cefditoren pivaloyloxymethyl ester ME 1207
Solubility Information DMSO : ≥ 100 mg/mL (161.10 mM)
SMILES O=C(C(N12)=C(/C=C\C3=C(C)N=CS3)CS[C@]2([H])[C@H](NC(/C(C4=CSC(N)=N4)=N\OC)=O)C1=O)OCOC(C(C)(C)C)=O
Molecular Weight (g/mol) 620.72
Formula Weight 620.72
Percent Purity 99.31%
Grade Research