missing translation for 'onlineSavingsMsg'
Learn More

Cav 2.2 blocker 1, MedChemExpress

Product Code. 16434898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434898 10 mg 10mg
16487647 5 mg 5mg
16461147 50 mg 50mg
16445676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16434898 Supplier Medchem Express Supplier No. HY119373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC50 of 1 nM.

Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC50 of 1 nM.

Specifications

Chemical Name or Material Cav 2.2 blocker 1
CAS 1567335-29-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H29ClN2O2
Quantity 10 mg
Solubility Information DMSO : 20.83 mg/mL (49.02 mM; Need ultrasonic)
SMILES COC(C=CC=C1)=C1N2C(C3=CC=C(Cl)C=C3)=CC(C4CC(C)(C)OC(C)(C)C4)=N2
Molecular Weight (g/mol) 424.96
Formula Weight 424.96
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.