missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoylmesaconine, MedChemExpress
Description
Benzoylmesaconine is the most abundant component of Wutou decoction, which is widely used in China because of its therapeutic effect on rheumatoid arthritis.
Specifications
Specifications
| Chemical Name or Material | Benzoylmesaconine |
| CAS | 63238-67-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H43NO10 |
| Quantity | 10 mg |
| Synonym | Mesaconine 14-benzoate |
| Solubility Information | DMSO : 100 mg/mL (169.59 mM; Need ultrasonic) |
| SMILES | CO[C@@H]1C2([C@@](C[C@@]3(O)[C@@H]4OC(C5=CC=CC=C5)=O)([H])[C@@]4([H])[C@](O)([C@@H](O)[C@@H]3OC)C6C2N(C)C7)[C@@]([C@H]6OC)([H])[C@@]7(COC)[C@H](O)C1 |
| Molecular Weight (g/mol) | 589.67 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?