missing translation for 'onlineSavingsMsg'
Learn More

Bay 65-1942 hydrochloride, MedChemExpress

Product Code. 16418158
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418158 10 mg 10mg
16440608 5 mg 5mg
16440356 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16418158 Supplier Medchem Express Supplier No. HY50948
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor.

Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor.

Specifications

Chemical Name or Material Bay 65-1942 hydrochloride
CAS 600734-06-3
Color White
Purity Grade Notes Research
Molecular Formula C22H26ClN3O4
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (115.76 mM; Need ultrasonic) ∣H2O : 2.17 mg/mL (5.02 mM; Need ultrasonic)
SMILES O=C1OCC2=C([C@H]3CNCCC3)C=C(C4=C(O)C=CC=C4OCC5CC5)N=C2N1.[H]Cl
Molecular Weight (g/mol) 431.91
Formula Weight 431.91
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.