missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan medoxomil, MedChemExpress

Product Code. 16466148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466148 10 mg 10mg
16488747 5 mg 5mg
16442287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466148 Supplier Medchem Express Supplier No. HY14736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Specifications

Chemical Name or Material Azilsartan medoxomil
CAS 863031-21-4
Color White
Purity Grade Notes Research
Molecular Formula C30H24N4O8
Quantity 10 mg
Synonym TAK-491
Solubility Information DMSO : 125 mg/mL (219.87 mM; Need ultrasonic)
SMILES O=C(C1=C2C(N=C(OCC)N2CC3=CC=C(C4=CC=CC=C4C5=NC(ON5)=O)C=C3)=CC=C1)OCC6=C(C)OC(O6)=O
Molecular Weight (g/mol) 568.53
Formula Weight 568.53
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.