missing translation for 'onlineSavingsMsg'
Learn More

Atropine methyl bromide, MedChemExpress

Product Code. 16464558
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16464558 10 mg 10mg
16490857 50 mg 50mg
16485406 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 16464558

Brand: Medchem Express HY112076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine.

Atropine methyl bromide, a muscarinic receptor (mAChR) antagonist, is a quaternary ammonium salt of atropine and a mydriatic for dilation of the pupil during ophthalmic examination. It is introduced for relieving pyloric spasm in infants for its highly polar nature. It penetrates less readily into the central nervous system than atropine.

Specifications

Chemical Name or Material Atropine methyl bromide
Physical Form Solid
CAS 2870-71-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H26BrNO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym Methylatropine bromide
Solubility Information DMSO : 150 mg/mL (390.31 mM; Need ultrasonic)
SMILES C[N+]1([C@H]2C[C@H](OC(C(C3=CC=CC=C3)CO)=O)C[C@@H]1CC2)C.[Br-]
Molecular Weight (g/mol) 384.31
Formula Weight 384.31
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.